Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGGTATCAGTGTCTGAACAGG[A/G]CCCACCTCAGCCCCAAGGCCACCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Zmat4 PubMed Links |
Zmat4 - zinc finger, matrin type 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277239.1 | Intron | NP_001264168.1 | ||||
NM_177086.4 | Intron | NP_796060.1 |