Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGACTGCTGGTGACTGACATGAG[C/T]GATGCTGAGCAATACAGAAGCTACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hat1 PubMed Links |
Hat1 - histone aminotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026115.4 | 1058 | Silent Mutation | NP_080391.2 |