Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATAAGCCTTTATCAAGAACAAA[A/G]CATAAATGGCAGATTCTTTACATGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh20 PubMed Links |
Cdh20 - cadherin 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011800.4 | Intron | NP_035930.1 |