Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGTTTGAAGGTATAGCTTTTT[A/C]TGAGAGTTTCATTCATGTGTATATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Frmd3 PubMed Links |
Frmd3 - FERM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163732.1 | Intron | NP_001157204.1 | ||||
NM_172869.4 | Intron | NP_766457.1 |