Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Glod4 PubMed Links |
Glod4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Nxn - nucleoredoxin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Rnmtl1 - RNA methyltransferase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_183263.5 | 645 | Missense Mutation | NP_899086.2 |