Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCATTGCACAGGGACTCCCAAGA[G/T]CGAAGAGACATCACTCACCACAGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Col6a1 PubMed Links |
Col6a1 - collagen, type VI, alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009933.4 | Intron | NP_034063.1 |