Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACTTCAAACCCCATTTATGCTT[A/C]ATTTCAAAGACATTTGTCTTTTTTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Auts2 PubMed Links |
Auts2 - autism susceptibility candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177047.3 | Intron | NP_796021.2 |