Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATAATGCAATTAGAAATGGGC[A/G]CTGTGCTTGGGTGGCATGATTGCTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mllt3 PubMed Links |
Mllt3 - myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286158.1 | Intron | NP_001273087.1 | ||||
NM_027326.3 | Intron | NP_081602.3 | ||||
NM_029931.3 | Intron | NP_084207.2 |