Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nsmaf PubMed Links |
Nsmaf - neutral sphingomyelinase (N-SMase) activation associated factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Sdcbp - syndecan binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098227.1 | Intron | NP_001091697.1 | ||||
NM_016807.2 | Intron | NP_058087.2 |