Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ap4m1 PubMed Links |
Ap4m1 - adaptor-related protein complex AP-4, mu 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021392.4 | Intron | NP_067367.3 |
Cnpy4 - canopy 4 homolog (zebrafish) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mcm7 - minichromosome maintenance deficient 7 (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Taf6 - TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |