Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGGGTGTTTTTGGTTTAAGAG[C/T]CAGATAGACACATACAGATGTATAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kctd16 PubMed Links |
Kctd16 - potassium channel tetramerisation domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026135.1 | Intron | NP_080411.1 |