Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCTGGAGCAGGGCATGCTAGTA[A/G]AGAGAAATTGTTCATCACTGGATCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Eci2 PubMed Links |
Eci2 - enoyl-Coenzyme A delta isomerase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110331.1 | Intron | NP_001103801.1 | ||||
NM_011868.3 | Intron | NP_035998.2 |