Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGCTGTGTCTTTATGCCTGCA[C/T]GGTGGCTTGTATTTGCTTTCTCTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rims2 PubMed Links |
Rims2 - regulating synaptic membrane exocytosis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256382.1 | Intron | NP_001243311.1 | ||||
NM_001256383.1 | Intron | NP_001243312.1 | ||||
NM_001256384.1 | Intron | NP_001243313.1 | ||||
NM_053271.2 | Intron | NP_444501.1 |