Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAGTCACCTGATGACTTTATTT[A/G]AAATAACAGTCCATTCATAGAAACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tshz2 PubMed Links |
Tshz2 - teashirt zinc finger family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080455.2 | Intron | NP_536703.2 |