Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTTGTTTCTAAAGAGCCCCACTT[A/C]ATGTCAACTCCTGTTTCTTTAACTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptprg PubMed Links |
Ptprg - protein tyrosine phosphatase, receptor type, G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008981.3 | 8622 | UTR 3 | NP_033007.2 |