Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATATATGCTCACTTTCAACCAA[G/T]GGCTTTGTGTTGCGAACAGTGGCCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fars2 PubMed Links |
Fars2 - phenylalanine-tRNA synthetase 2 (mitochondrial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039189.2 | Intron | NP_001034278.1 | ||||
NM_024274.3 | Intron | NP_077236.1 |