Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGCATGTGGCATGTTCATGGCA[C/T]GTGGCACAGACATCTACATCTGAGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Susd1 PubMed Links |
Susd1 - sushi domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163288.2 | 448 | Silent Mutation | NP_001156760.2 |