Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATAGTTTTTGTCCAAGAGTAATC[C/T]GATGATGGGCCTAGAAAGGTCTACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gng2 PubMed Links |
Gng2 - guanine nucleotide binding protein (G protein), gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038637.1 | Intron | NP_001033726.1 | ||||
NM_001285908.1 | Intron | NP_001272837.1 | ||||
NM_001285909.1 | Intron | NP_001272838.1 | ||||
NM_001285910.1 | Intron | NP_001272839.1 | ||||
NM_001285911.1 | Intron | NP_001272840.1 | ||||
NM_010315.4 | Intron | NP_034445.1 |