Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGCTGCGCTCTCCCCCAGAGCC[A/G]GCCAGGACACAGCTGGCTTCTCCAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Musk PubMed Links |
Musk - muscle, skeletal, receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037127.2 | Intron | NP_001032204.2 | ||||
NM_001037128.1 | Intron | NP_001032205.1 | ||||
NM_001037129.1 | Intron | NP_001032206.1 | ||||
NM_001037130.1 | Intron | NP_001032207.1 | ||||
NM_001165996.1 | Intron | NP_001159468.1 | ||||
NM_010944.2 | Intron | NP_035074.2 |