Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTAAGGGCTGTGTCACAGATTA[C/T]TTCCTTAAAACAGATTTCTCTTAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arid1b PubMed Links |
Arid1b - AT rich interactive domain 1B (SWI-like) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085355.1 | Intron | NP_001078824.1 |