Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAGGTCACCTAAGGCCCTAACA[A/C]TCACTGGGGAAAGGAGTACCCCTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Spata16 PubMed Links |
Spata16 - spermatogenesis associated 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027583.3 | Intron | NP_081859.1 | ||||
NM_029150.4 | Intron | NP_083426.1 |