Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm26881 PubMed Links |
Gm26881 - predicted gene, 26881 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Unc13b - unc-13 homolog B (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081413.1 | Intron | NP_001074882.1 | ||||
NM_021468.2 | Intron | NP_067443.2 |