Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTAAATGACTAGCTGTGACAG[A/T]ACTACCATCAGCAAAGCCTTGCAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slamf6 PubMed Links |
Slamf6 - SLAM family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030710.2 | Intron | NP_109635.1 |