Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTATCCATCAGTCATCTCAAG[T/C]AGAGATATGCAGGCCTGAAGGACAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Syn3 PubMed Links |
Syn3 - synapsin III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164495.1 | Intron | NP_001157967.1 | ||||
NM_013722.3 | Intron | NP_038750.2 |