Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fam126b PubMed Links |
Fam126b - family with sequence similarity 126, member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172513.3 | Intron | NP_766101.3 |
Ndufb3 - NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025597.2 | Intron | NP_079873.1 |