Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCACTGCTGGAGGGGCGCAGATT[C/T]CCTTCAAAACAGGTGAGGTGATGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myo10 PubMed Links |
Myo10 - myosin X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019472.2 | 1508 | Missense Mutation | NP_062345.2 |