Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGGTTTAACTCGAGTCAAAGTA[C/T]TTGCCTGAATCTTGATTACTGACTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tmc1 PubMed Links |
Tmc1 - transmembrane channel-like gene family 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028953.2 | Intron | NP_083229.1 |