Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTAGCTAACTGGAACTGCAGAT[A/C]TGAGAGTGGCCTGGTTCCTTTCTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Xrcc5 PubMed Links |
Xrcc5 - X-ray repair complementing defective repair in Chinese hamster cells 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009533.2 | Intron | NP_033559.2 |