Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAAATTCCTTAAACCAAGACTA[C/T]GGTTAGGGGTCCATAATGCAAGGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Osbpl1a PubMed Links |
Osbpl1a - oxysterol binding protein-like 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252489.1 | Intron | NP_001239418.1 | ||||
NM_001252490.1 | Intron | NP_001239419.1 | ||||
NM_001252491.1 | Intron | NP_001239420.1 | ||||
NM_001252492.1 | Intron | NP_001239421.1 | ||||
NM_001252493.1 | Intron | NP_001239422.1 | ||||
NM_207530.3 | Intron | NP_997413.2 |