Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATTAATTATCTTTACTTCATCT[C/T]GAGGATTTTGAAATTAGGGAATTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Acaca PubMed Links |
Acaca - acetyl-Coenzyme A carboxylase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133360.2 | Intron | NP_579938.2 |