Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACATATACATTCCGAATTTCGTC[T/A]CGACTGGCATTTACCCACTCTGATA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700027A15Rik PubMed Links |
1700027A15Rik - RIKEN cDNA 1700027A15 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tnp1 - transition protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009407.2 | 342 | UTR 3 | NP_033433.1 |