Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACTGGCCTGTTTTTTCCTTTC[C/T]TTTCTTTTCTTTTCTTTTCTTTTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ptprm PubMed Links |
Ptprm - protein tyrosine phosphatase, receptor type, M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008984.2 | Intron | NP_033010.2 |