Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTTCCTTTTTCCTAAAACACA[C/T]GCAAATCAAAACTAGAGTCTAAACC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fstl4 PubMed Links |
Fstl4 - follistatin-like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177059.3 | Intron | NP_796033.2 |