Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bcs1l PubMed Links |
Bcs1l - BCS1-like (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Rnf25 - ring finger protein 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Zfp142 - zinc finger protein 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029888.2 | Intron | NP_084164.1 |