Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCTGTGGTTCCTGGTCGTGACC[A/G]CTGGGATCCCTCTCGTAGCTCTCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pgm5 PubMed Links |
Pgm5 - phosphoglucomutase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175013.2 | 1646 | Silent Mutation | NP_778178.3 |