Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdk17 PubMed Links |
Cdk17 - cyclin-dependent kinase 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146239.2 | Intron | NP_666351.2 |
Elk3 - ELK3, member of ETS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |