Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Armc7 PubMed Links |
Armc7 - armadillo repeat containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Hn1 - hematological and neurological expressed sequence 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Nt5c - 5',3'-nucleotidase, cytosolic | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015807.1 | 85 | Silent Mutation | NP_056622.1 |