Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCTGGGGCGGGAATGGGGGACA[A/C]GCCTTAAGCGGAAATAAAAATAGCG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Strn PubMed Links |
Strn - striatin, calmodulin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011500.2 | Intron | NP_035630.2 |