Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hddc2 PubMed Links |
Hddc2 - HD domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027168.2 | Intron | NP_081444.1 |
Tpd52l1 - tumor protein D52-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |