Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCGAACTTGTTAGGAAAACAAC[A/G]AGAACCACTCACCTTAAGGTATGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fut8 PubMed Links |
Fut8 - fucosyltransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252614.1 | Intron | NP_001239543.1 | ||||
NM_001252615.1 | Intron | NP_001239544.1 | ||||
NM_001252616.1 | Intron | NP_001239545.1 | ||||
NM_016893.5 | Intron | NP_058589.2 |