Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCCCTCTAATACAGATGGCAC[A/G]ATGAACCATCAGAAGGAAAATGGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Itgb6 PubMed Links |
Itgb6 - integrin beta 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159564.1 | Intron | NP_001153036.1 | ||||
NM_021359.3 | Intron | NP_067334.1 |