Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mir1943 PubMed Links |
Mir1943 - microRNA 1943 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Tmem184b - transmembrane protein 184b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253817.1 | 362 | Silent Mutation | NP_001240746.1 | |||
NM_001253819.1 | 362 | Silent Mutation | NP_001240748.1 | |||
NM_001253820.1 | 362 | Intron | NP_001240749.1 | |||
NM_172608.1 | 362 | Silent Mutation | NP_766196.1 |