Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTACTTCCTAGCATGCACCTAGC[A/G]TAAACTGCGGCAAATTATCAACGAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc44a1 PubMed Links |
Slc44a1 - solute carrier family 44, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159633.1 | Intron | NP_001153105.1 | ||||
NM_133891.3 | Intron | NP_598652.3 |