Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAGAAAGTTACAGCTTTTTTCT[A/G]AAGTTTAGGATTTGTAGTGCTGTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Melk PubMed Links |
Melk - maternal embryonic leucine zipper kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010790.2 | Intron | NP_034920.2 |