Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fbxo38 PubMed Links |
Fbxo38 - F-box protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_134136.3 | 4342 | UTR 3 | NP_598897.2 |
Htr4 - 5 hydroxytryptamine (serotonin) receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |