Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAACTCGAAAGAACAGGACTG[C/T]CCTGCTGTCTGAGGGGACCCTGGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
B3galt5 PubMed Links |
B3galt5 - UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122993.1 | 1506 | Silent Mutation | NP_001116465.1 | |||
NM_033149.3 | 1506 | Silent Mutation | NP_149161.1 |