Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Crygs PubMed Links |
Crygs - crystallin, gamma S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009967.2 | 436 | Silent Mutation | NP_034097.1 |
Tbccd1 - TBCC domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |