Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTTGTTTCCCACAGGCTTTGCA[C/T]GGTGTCTTGACTTCTCAAACTACAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
L3mbtl4 PubMed Links |
L3mbtl4 - l(3)mbt-like 4 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177278.5 | Intron | NP_796252.2 |