Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATCTGTATTGAACAGGCCCAGG[C/G]CTTGTTATCCTTCTCTAAACAGTAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rgs6 PubMed Links |
Rgs6 - regulator of G-protein signaling 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282061.1 | Intron | NP_001268990.1 | ||||
NM_015812.3 | Intron | NP_056627.1 |