Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTTTGGGAAGGAGTAAGAGGAA[A/G]CACTGGCTGCCACTTCTATCTTTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pax8 PubMed Links |
Pax8 - paired box 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011040.4 | Intron | NP_035170.1 |